missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

cis-5-Norbornene-endo-2,3-dicarboxylic anhydride, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2495106

 View more versions of this product

Catalog No. AAB2495106



Description

Description

cis-5-Norbornene-endo-2,3-dicarboxylic anhydride is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-5-Norbornene-endo-2,3-dicarboxylic anhydride is used as an organic chemical synthesis intermediate.

Solubility
It decomposes in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents.
Specifications

Specifications

cis-5-Norbornene-endo-2,3-dicarboxylic anhydride
164°C to 167°C
C9H8O3
5 g
Moisture sensitive
cis-5-norbornene-endo-2,3-dicarboxylic anhydride
KNDQHSIWLOJIGP-UHFFFAOYNA-N
4-oxatricyclo[5.2.1.0²,⁶]dec-8-ene-3,5-dione
126963424
97%
129-64-6
1.417
MFCD00151106,MFCD00069013,MFCD00630580,MFCD00151106
83657
14,1796
It decomposes in water.
O=C1OC(=O)C2C3CC(C=C3)C12
164.16
164.16

Chemical Identifiers

129-64-6
164.16
KNDQHSIWLOJIGP-UHFFFAOYNA-N
126963424
O=C1OC(=O)C2C3CC(C=C3)C12
C9H8O3
MFCD00151106,MFCD00069013,MFCD00630580,MFCD00151106
cis-5-norbornene-endo-2,3-dicarboxylic anhydride
4-oxatricyclo[5.2.1.0²,⁶]dec-8-ene-3,5-dione
Safety and Handling

Safety and Handling

GHS H Statement
H334-H318-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
May cause an allergic skin reaction.

P261-P272-P280-P285-P302+P352-P304+P341-P305+P351+P338-P310-P333+P313-P342+P311-P363-P501c

H317-H318-H334

EINECSNumber : 204-957-7

RTECSNumber : DT5600000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only