missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

cis-4-(tert-Butoxycarbonylamino)cyclohexanecarboxylic Acid, TCI America™

$166.53 - $511.81

Chemical Identifiers

CAS 53292-90-3
Molecular Formula C12H21NO4
Molecular Weight (g/mol) 243.303
MDL Number MFCD01862294
InChI Key KXMRDHPZQHAXML-UHFFFAOYSA-N
Synonym trans-4-boc-amino cyclohexanecarboxylic acid, cis-4-boc-amino cyclohexanecarboxylic acid, 4-boc-amino cyclohexanecarboxylic acid, cis-4-tert-butoxycarbonylamino cyclohexanecarboxylic acid, trans-4-tert-butoxycarbonylamino cyclohexanecarboxylic acid, boc-trans-4-aminocyclohexanecarboxylic acid, boc-1,4-trans-achc-oh, cis-4-tert-butoxycarbonyl amino cyclohexanecarboxylic acid, 4-tert-butoxycarbonyl amino cyclohexanecarboxylic acid, 4-tert-butoxycarbonylamino cyclohexanecarboxylic acid
PubChem CID 2755996
IUPAC Name 4-[(2-methylpropan-2-yl)oxycarbonylamino]cyclohexane-1-carboxylic acid
SMILES CC(C)(C)OC(=O)NC1CCC(CC1)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B32495G View Documents TCI America
B32495G
5g Each for $166.53
 
B324925G View Documents TCI America
B324925G
25g Each for $511.81
 
Specifications

Chemical Identifiers

53292-90-3
243.303
KXMRDHPZQHAXML-UHFFFAOYSA-N
2755996
CC(C)(C)OC(=O)NC1CCC(CC1)C(=O)O
C12H21NO4
MFCD01862294
trans-4-boc-amino cyclohexanecarboxylic acid, cis-4-boc-amino cyclohexanecarboxylic acid, 4-boc-amino cyclohexanecarboxylic acid, cis-4-tert-butoxycarbonylamino cyclohexanecarboxylic acid, trans-4-tert-butoxycarbonylamino cyclohexanecarboxylic acid, boc-trans-4-aminocyclohexanecarboxylic acid, boc-1,4-trans-achc-oh, cis-4-tert-butoxycarbonyl amino cyclohexanecarboxylic acid, 4-tert-butoxycarbonyl amino cyclohexanecarboxylic acid, 4-tert-butoxycarbonylamino cyclohexanecarboxylic acid
4-[(2-methylpropan-2-yl)oxycarbonylamino]cyclohexane-1-carboxylic acid

Specifications

White-Yellow
5g
C12H21NO4
trans-4-boc-amino cyclohexanecarboxylic acid, cis-4-boc-amino cyclohexanecarboxylic acid, 4-boc-amino cyclohexanecarboxylic acid, cis-4-tert-butoxycarbonylamino cyclohexanecarboxylic acid, trans-4-tert-butoxycarbonylamino cyclohexanecarboxylic acid, boc-trans-4-aminocyclohexanecarboxylic acid, boc-1,4-trans-achc-oh, cis-4-tert-butoxycarbonyl amino cyclohexanecarboxylic acid, 4-tert-butoxycarbonyl amino cyclohexanecarboxylic acid, 4-tert-butoxycarbonylamino cyclohexanecarboxylic acid
CC(C)(C)OC(=O)NC1CCC(CC1)C(=O)O
243.303
243.30
≥98.0% (GC,T)
169°C
53292-90-3
MFCD01862294
KXMRDHPZQHAXML-UHFFFAOYSA-N
4-[(2-methylpropan-2-yl)oxycarbonylamino]cyclohexane-1-carboxylic acid
2755996
Crystal-Powder at 20°C
cis-4-(tert-Butoxycarbonylamino)cyclohexanecarboxylic Acid
Safety and Handling

Safety and Handling

TSCA : No