missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

cis-4-Nitrostilbene, TCI America™

$238.34 - $711.95

Chemical Identifiers

CAS 6624-53-9
Molecular Formula C14H11NO2
Molecular Weight (g/mol) 225.247
InChI Key ZISCOWXWCHUSMH-SREVYHEPSA-N
Synonym (Z)-4-Nitrostilbene
PubChem CID 1551577
IUPAC Name 1-nitro-4-[(Z)-2-phenylethenyl]benzene
SMILES C1=CC=C(C=C1)C=CC2=CC=C(C=C2)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N10851G View Documents TCI America
N10851G
1g Each for $711.95
 
N1085200MG View Documents TCI America
N1085200MG
200mg Each for $238.34
 
Specifications

Chemical Identifiers

6624-53-9
225.247
(Z)-4-Nitrostilbene
1-nitro-4-[(Z)-2-phenylethenyl]benzene
C14H11NO2
ZISCOWXWCHUSMH-SREVYHEPSA-N
1551577
C1=CC=C(C=C1)C=CC2=CC=C(C=C2)[N+](=O)[O-]

Specifications

Undesignated
1g
C14H11NO2
ZISCOWXWCHUSMH-SREVYHEPSA-N
1-nitro-4-[(Z)-2-phenylethenyl]benzene
1551577
Crystal-Powder at 20°C
cis-4-Nitrostilbene
63°C
6624-53-9
(Z)-4-Nitrostilbene
C1=CC=C(C=C1)C=CC2=CC=C(C=C2)[N+](=O)[O-]
225.247
225.25
≥97.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No