missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

cis-4-Methylcyclohexanol 98.0+%, TCI America™

Manufacturer:  TCI America M03531ML

 View more versions of this product

Catalog No. M03531ML



Specifications

Specifications

cis-4-Methylcyclohexanol
Colorless
7731-28-4
MFCD00064170
MQWCXKGKQLNYQG-UHFFFAOYSA-N
4-methylcyclohexan-1-ol
11524
Clear Liquid at 20°C
171°C
1mL
C7H14O
4-methylcyclohexanol, trans-4-methylcyclohexanol, cis-4-methylcyclohexanol, cyclohexanol, 4-methyl, 1-methyl-4-cyclohexanol, cyclohexanol, 4-methyl-, trans, methyl cyclohexanol, p-methylcyclohexanol, cyclohexanol, 4-methyl-, cis, trans 4-methyl cyclohexanol
CC1CCC(CC1)O
114.188
114.19
≥98.0% (GC)

Chemical Identifiers

7731-28-4
114.188
MQWCXKGKQLNYQG-UHFFFAOYSA-N
11524
CC1CCC(CC1)O
C7H14O
MFCD00064170
4-methylcyclohexanol, trans-4-methylcyclohexanol, cis-4-methylcyclohexanol, cyclohexanol, 4-methyl, 1-methyl-4-cyclohexanol, cyclohexanol, 4-methyl-, trans, methyl cyclohexanol, p-methylcyclohexanol, cyclohexanol, 4-methyl-, cis, trans 4-methyl cyclohexanol
4-methylcyclohexan-1-ol
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2301

TSCA : No

Documents