missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

cis-4-(Hydroxymethyl)cyclohexanecarboxylic Acid, TCI America™

$325.42 - $1,138.22

Chemical Identifiers

CAS 73094-35-6
Molecular Formula C8H14O3
Molecular Weight (g/mol) 158.197
InChI Key VQMIUUBKKPIDBN-UHFFFAOYSA-N
PubChem CID 202819
IUPAC Name 4-(hydroxymethyl)cyclohexane-1-carboxylic acid
SMILES C1CC(CCC1CO)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H12425G View Documents TCI America
H12425G
5g Each for $325.42
 
H124225G View Documents TCI America
H124225G
25g Each for $1,138.22
 
Specifications

Chemical Identifiers

73094-35-6
158.197
202819
C1CC(CCC1CO)C(=O)O
C8H14O3
VQMIUUBKKPIDBN-UHFFFAOYSA-N
4-(hydroxymethyl)cyclohexane-1-carboxylic acid

Specifications

White-Yellow
5g
C8H14O3
C1CC(CCC1CO)C(=O)O
158.197
158.20
≥98.0% (GC)
120°C
73094-35-6
VQMIUUBKKPIDBN-UHFFFAOYSA-N
4-(hydroxymethyl)cyclohexane-1-carboxylic acid
202819
Crystal-Powder at 20°C
cis-4-(Hydroxymethyl)cyclohexanecarboxylic Acid
Safety and Handling

Safety and Handling

TSCA : No