missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
cis-4-(Hydroxymethyl)cyclohexanecarboxylic Acid 98.0+%, TCI America™
Supplier: TCI America H124225G
Specifications
cis-4-(Hydroxymethyl)cyclohexanecarboxylic Acid | |
120°C | |
C8H14O3 | |
VQMIUUBKKPIDBN-UHFFFAOYSA-N | |
4-(hydroxymethyl)cyclohexane-1-carboxylic acid | |
202819 | |
≥98.0% (GC) |
73094-35-6 | |
White-Yellow | |
25 g | |
C1CC(CCC1CO)C(=O)O | |
158.197 | |
158.20 | |
Crystalline Powder |
Chemical Identifiers
73094-35-6 | |
158.197 | |
202819 | |
C1CC(CCC1CO)C(=O)O |
C8H14O3 | |
VQMIUUBKKPIDBN-UHFFFAOYSA-N | |
4-(hydroxymethyl)cyclohexane-1-carboxylic acid |
Safety and Handling
TSCA : No