missing translation for 'onlineSavingsMsg'
Learn More

cis-4-Hydroxycyclohexylamine hydrochloride, 95%

CAS: 56239-26-0 | C6H14ClNO | 151.63 g/mol

$163.82 - $515.47

Chemical Identifiers

CAS 56239-26-0
Molecular Formula C6H14ClNO
Molecular Weight (g/mol) 151.63
MDL Number MFCD00012566,MFCD07366531,MFCD06410647
InChI Key RKTQEVMZBCBOSB-UHFFFAOYSA-N
Synonym trans-4-aminocyclohexanol hydrochloride, cis-4-aminocyclohexanol hydrochloride, 4-aminocyclohexanol hydrochloride, cis-4-aminocyclohexanol hcl, 4-aminocyclohexan-1-ol hydrochloride, cis-4-amino-cyclohexanol hydrochloride, 4-amino-cyclohexanol hydrochloride, 1r,4r-4-aminocyclohexan-1-ol hydrochloride, cyclohexanol, 4-amino-, hydrochloride, trans-4-aminocyclohexanol hcl
PubChem CID 522619
IUPAC Name 4-aminocyclohexan-1-ol;hydrochloride
SMILES [H+].[Cl-].NC1CCC(O)CC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH66110MD
View Documents
Thermo Scientific Chemicals
H66110MD
250 mg
Each for $163.82
Only null left
 
AAH6611003
View Documents
Thermo Scientific Chemicals
H6611003
1 g
Each for $515.47
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

56239-26-0
151.63
RKTQEVMZBCBOSB-UHFFFAOYSA-N
522619
[H+].[Cl-].NC1CCC(O)CC1
C6H14ClNO
MFCD00012566,MFCD07366531,MFCD06410647
trans-4-aminocyclohexanol hydrochloride, cis-4-aminocyclohexanol hydrochloride, 4-aminocyclohexanol hydrochloride, cis-4-aminocyclohexanol hcl, 4-aminocyclohexan-1-ol hydrochloride, cis-4-amino-cyclohexanol hydrochloride, 4-amino-cyclohexanol hydrochloride, 1r,4r-4-aminocyclohexan-1-ol hydrochloride, cyclohexanol, 4-amino-, hydrochloride, trans-4-aminocyclohexanol hcl
4-aminocyclohexan-1-ol;hydrochloride

Specifications

56239-26-0
C6H14ClNO
250 mg
Moisture sensitive
RKTQEVMZBCBOSB-UHFFFAOYSA-N
4-aminocyclohexan-1-ol;hydrochloride
522619
95%
cis-4-Hydroxycyclohexylamine hydrochloride
193°C to 200°C
MFCD00012566,MFCD07366531,MFCD06410647
3909295
trans-4-aminocyclohexanol hydrochloride, cis-4-aminocyclohexanol hydrochloride, 4-aminocyclohexanol hydrochloride, cis-4-aminocyclohexanol hcl, 4-aminocyclohexan-1-ol hydrochloride, cis-4-amino-cyclohexanol hydrochloride, 4-amino-cyclohexanol hydrochloride, 1r,4r-4-aminocyclohexan-1-ol hydrochloride, cyclohexanol, 4-amino-, hydrochloride, trans-4-aminocyclohexanol hcl
[H+].[Cl-].NC1CCC(O)CC1
151.63
151.63
Solid
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only