missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
cis-4-Hydroxycyclohexanecarboxylic Acid, TCI America™
$235.28 - $727.23
Chemical Identifiers
CAS | 3685-22-1 |
---|---|
Molecular Formula | C7H12O3 |
Molecular Weight (g/mol) | 144.17 |
MDL Number | MFCD04038017 |
InChI Key | HCFRWBBJISAZNK-UHFFFAOYSA-N |
PubChem CID | 151138 |
IUPAC Name | 4-hydroxycyclohexane-1-carboxylic acid |
SMILES | C1CC(CCC1C(=O)O)O |
Chemical Identifiers
3685-22-1 | |
144.17 | |
HCFRWBBJISAZNK-UHFFFAOYSA-N | |
4-hydroxycyclohexane-1-carboxylic acid |
C7H12O3 | |
MFCD04038017 | |
151138 | |
C1CC(CCC1C(=O)O)O |
Specifications
White | |
25g | |
C7H12O3 | |
HCFRWBBJISAZNK-UHFFFAOYSA-N | |
4-hydroxycyclohexane-1-carboxylic acid | |
151138 | |
Crystal-Powder at 20°C | |
cis-4-Hydroxycyclohexanecarboxylic Acid |
150°C | |
3685-22-1 | |
MFCD04038017 | |
C1CC(CCC1C(=O)O)O | |
144.17 | |
144.17 | |
≥98.0% (GC,T) |
Safety and Handling
TSCA : No