Please login to your online account to display your discounted pricing

cis-4-Hydroxy-L-proline, 99%, ACROS Organics™


Quantity 50mg
Packaging Glass bottle

Manufacturer: fisher scientific acros  204920500

 View more versions of this product

Catalog No. AC204920500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Formula Weight 131.13
Physical Form Powder
Quantity 50mg
Color White to beige
Merck Index 15, 4878
Beilstein 22, I, 546
TSCA TSCA
EINECS Number 210-542-1
Specific Rotation Condition −59.00 (20.00°C c=2,H2O)
Infrared Spectrum Authentic
Specific Rotation -58° to -60° (20°C, 589nm) (c=2, H2O)
Packaging Glass bottle
Synonym (2S, 4S)-(-)-4-Hydroxy-2-pyrrolinecarboxylic acid
Name Note 99%
Molecular Formula C5H9NO3
MDL Number MFCD00064319
Chemical Name or Material cis-4-Hydroxy-L-proline
CAS 618-27-9
Assay Percent Range 99%

Specifications

Chemical Name or Material cis-4-Hydroxy-L-proline
Name Note 99%
CAS 618-27-9
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C5H9NO3
Formula Weight 131.13
MDL Number MFCD00064319
Synonym (2S, 4S)-(-)-4-Hydroxy-2-pyrrolinecarboxylic acid