Please login to your online account to display your discounted pricing

cis-4-Hydroxy-D-proline, 99%, ACROS Organics™


Quantity 250mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC204912500

 View more versions of this product

Catalog No. AC204912500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Formula Weight 131.13
Physical Form Powder
Quantity 250mg
Color White
Specific Rotation Condition +58° (24°C c=2,water)
Beilstein 22, I, 546
Merck Index 10, 4762
Specific Rotation +56° to +60° (24°C, 589nm) (c=2, water)
Chemical Name or Material cis-4-Hydroxy-D-proline
Assay Percent Range 99%
CAS 2584-71-6
Molecular Formula C5H9NO3
MDL Number MFCD00005252
Packaging Glass bottle
Infrared Spectrum Authentic

Specifications

Chemical Name or Material cis-4-Hydroxy-D-proline
CAS 2584-71-6
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C5H9NO3
Formula Weight 131.13
MDL Number MFCD00005252