missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

cis-4-Hexen-1-ol, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC439740050

 View more versions of this product

Catalog No. AC439740050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

cis-4-Hexen-1-ol
65°C
Glass bottle
70°C (28.0mmHg)
5mL
96% min. (GC)
4-hexen-1-ol, z, 4z-4-hexen-1-ol #, 4z-hex-4-en-1-ol, cis-1-hydroxy-4-hexene, cis-4-hexen-1-ol, cis-4-hexene-1-ol, e-4-hexenol, unii-780wma6j84, z-4-hexen-1-ol, z-hex-4-en-1-ol
CC=CCCCO
100.16
100.16
97%
0.8570g/mL
Authentic
0.857
Colorless
928-91-6
C6H12O
VTIODUHBZHNXFP-IHWYPQMZSA-N
(Z)-hex-4-en-1-ol
5365589
Liquid

Chemical Identifiers

928-91-6
100.16
4-hexen-1-ol, z, 4z-4-hexen-1-ol #, 4z-hex-4-en-1-ol, cis-1-hydroxy-4-hexene, cis-4-hexen-1-ol, cis-4-hexene-1-ol, e-4-hexenol, unii-780wma6j84, z-4-hexen-1-ol, z-hex-4-en-1-ol
(Z)-hex-4-en-1-ol
C6H12O
VTIODUHBZHNXFP-IHWYPQMZSA-N
5365589
CC=CCCCO
Safety and Handling

Safety and Handling

GHS H Statement:

GHS P Statement:
Do not get in eyes, on skin, or on clothing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 213-187-

SDS Documents