Please login to your online account to display your discounted pricing

cis-4-Hexen-1-ol, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material cis-4-Hexen-1-ol
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H12O
Formula Weight 100.16
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC439740050

  SDS
thermo fisher scientific
AC439740050
Glass bottle 5mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC439740250

  SDS
thermo fisher scientific
AC439740250
Glass bottle 25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Formula Weight 100.16
Assay ≥96.0%
Quantity 5mL
Color Clear colorless
Density 0.857g/mL
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Do not get in eyes, on skin, or on clothing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Molecular Formula C6H12O
Chemical Name or Material cis-4-Hexen-1-ol
Assay Percent Range 97%
CAS 928-91-6
Infrared Spectrum Authentic
Safety and Handling

GHS P Statement:
Do not get in eyes, on skin, or on clothing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement: