missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

cis-4-Hepten-1-ol, TCI America™

$132.92 - $429.31

Chemical Identifiers

CAS 6191-71-5
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.188
MDL Number MFCD00015575
InChI Key CUKAXHVLXKIPKF-ARJAWSKDSA-N
Synonym cis-4-hepten-1-ol, z-4-hepten-1-ol, z-hept-4-en-1-ol, cis-hept-4-enol, unii-802k6ztp9q, 4z-hept-4-en-1-ol, 4-hepten-1-ol, cis, 4z-4-hepten-1-ol, 4-hepten-1-ol, 4z, 4-hepten-1-ol, z
PubChem CID 5367536
IUPAC Name (Z)-hept-4-en-1-ol
SMILES CCC=CCCCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H083325ML View Documents TCI America
H083325ML
25mL Each for $429.31
 
H08335ML View Documents TCI America
H08335ML
5mL Each for $132.92
 
Specifications

Chemical Identifiers

6191-71-5
114.188
CUKAXHVLXKIPKF-ARJAWSKDSA-N
5367536
CCC=CCCCO
C7H14O
MFCD00015575
cis-4-hepten-1-ol, z-4-hepten-1-ol, z-hept-4-en-1-ol, cis-hept-4-enol, unii-802k6ztp9q, 4z-hept-4-en-1-ol, 4-hepten-1-ol, cis, 4z-4-hepten-1-ol, 4-hepten-1-ol, 4z, 4-hepten-1-ol, z
(Z)-hept-4-en-1-ol

Specifications

85°C
25mL
C7H14O
cis-4-hepten-1-ol, z-4-hepten-1-ol, z-hept-4-en-1-ol, cis-hept-4-enol, unii-802k6ztp9q, 4z-hept-4-en-1-ol, 4-hepten-1-ol, cis, 4z-4-hepten-1-ol, 4-hepten-1-ol, 4z, 4-hepten-1-ol, z
CCC=CCCCO
114.188
114.19
≥95.0% (GC)
Colorless
6191-71-5
MFCD00015575
CUKAXHVLXKIPKF-ARJAWSKDSA-N
(Z)-hept-4-en-1-ol
5367536
Clear Liquid at 20°C
cis-4-Hepten-1-ol
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2393

TSCA : Yes