Please login to your online account to display your discounted pricing

Alfa Aesar™ cis-4-Hepten-1-ol, 97%

$52.88 - $216.97

Chemical Identifiers

Chemical Name or Material cis-4-Hepten-1-ol
CAS 6191-71-5
Molecular Formula C7H14O
Formula Weight 114.18
MDL Number MFCD00015575
Beilstein 1840300
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1659603

 
alfa aesar™
A1659603
1g Each for $52.88

AAA1659606

 
alfa aesar™
A1659606
5g Each for $216.97
Description & Specifications

Specifications

Formula Weight 114.18
Assay 97%
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C7H14O
MDL Number MFCD00015575
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Flash Point 73°C (163°F)
Beilstein 1840300
Quantity 1g
Boiling Point 60°C (24mmHg)
TSCA TSCA
CAS 6191-71-5
Chemical Name or Material cis-4-Hepten-1-ol

Applications
cis-4-Hepten-1-ol is used for green topnotes. It is used as a flavoring agent for strawberry, tomato, dry, hay notes of mimosa and tomato vine. It also acts as stabilizers, surfactants and emulsifiers.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.