Please login to your online account to display your discounted pricing

Alfa Aesar™ cis-4-Cyclohexene-1,2-dicarboxylic anhydride, 95%

$35.72 - $35.72

Chemical Identifiers

Chemical Name or Material cis-4-Cyclohexene-1,2-dicarboxylic anhydride
CAS 935-79-5
Molecular Formula C8H8O3
Formula Weight 152.15
MDL Number MFCD00005916
Beilstein 82341
Synonym cis-1,2,3,6-Tetrahydrophthalic anhydride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2143130

 
alfa aesar™
B2143130
250g Each for $35.72
Description & Specifications

Specifications

Formula Weight 152.15
Chemical Name or Material cis-4-Cyclohexene-1,2-dicarboxylic anhydride
TSCA TSCA
Assay 95%
Molecular Formula C8H8O3
MDL Number MFCD00005916
CAS 935-79-5
Beilstein 82341
Synonym cis-1,2,3,6-Tetrahydrophthalic anhydride
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H334-H318-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
May cause an allergic skin reaction.
Melting Point 97° to 103°C
Quantity 250g
Health Hazard 3 GHS P Statement
P284a-P261-P280-P305+P351+P338-P310-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number GW5775000
Safety and Handling

Danger

GHS P Statement
P284a-P261-P280-P305+P351+P338-P310-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H334-H318-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
May cause an allergic skin reaction.