missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ cis-4-(Boc-amino)cyclohexanecarboxaldehyde, 97%

$66.62 - $791.11

Chemical Identifiers

CAS 181308-56-5
Molecular Formula C12H21NO3
Molecular Weight (g/mol) 227.304
MDL Number MFCD03844599
InChI Key GPDBIGSFXXKWQR-UHFFFAOYSA-N
Synonym tert-butyl trans-4-formylcyclohexylcarbamate, tert-butyl cis-4-formylcyclohexylcarbamate, trans-boc-4-aminocyclohexanecarbaldehyde, tert-butyl n-4-formylcyclohexyl carbamate, trans-4-formyl-cyclohexyl-carbamic acid tert-butyl ester, tert-butyl trans-4-formylcyclohexyl carbamate, tert-butyl 4-formylcyclohexyl carbamate, trans-tert-butyl 4-formylcyclohexyl carbamate, tert-butyl 1r,4r-4-formylcyclohexylcarbamate, cis-4-boc-amino cyclohexanecarboxaldehyde
PubChem CID 11020615
IUPAC Name tert-butyl N-(4-formylcyclohexyl)carbamate
SMILES CC(C)(C)OC(=O)NC1CCC(CC1)C=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62871MD View Documents Alfa Aesar™
H62871MD
250mg Each for $66.62
 
AAH6287103 View Documents Alfa Aesar™
H6287103
1g Each for $198.93
 
AAH6287106 View Documents Alfa Aesar™
H6287106
5g Each for $791.11
 
Description

Description

Applications
cis-4-(Boc-amino)cyclohexanecarboxaldehyde is used as a intermediate for chemical research.

Notes
Keep container tightly closed. Store away from oxidizing agent.
Specifications

Chemical Identifiers

181308-56-5
227.304
GPDBIGSFXXKWQR-UHFFFAOYSA-N
11020615
CC(C)(C)OC(=O)NC1CCC(CC1)C=O
C12H21NO3
MFCD03844599
tert-butyl trans-4-formylcyclohexylcarbamate, tert-butyl cis-4-formylcyclohexylcarbamate, trans-boc-4-aminocyclohexanecarbaldehyde, tert-butyl n-4-formylcyclohexyl carbamate, trans-4-formyl-cyclohexyl-carbamic acid tert-butyl ester, tert-butyl trans-4-formylcyclohexyl carbamate, tert-butyl 4-formylcyclohexyl carbamate, trans-tert-butyl 4-formylcyclohexyl carbamate, tert-butyl 1r,4r-4-formylcyclohexylcarbamate, cis-4-boc-amino cyclohexanecarboxaldehyde
tert-butyl N-(4-formylcyclohexyl)carbamate

Specifications

250mg
C12H21NO3
tert-butyl trans-4-formylcyclohexylcarbamate, tert-butyl cis-4-formylcyclohexylcarbamate, trans-boc-4-aminocyclohexanecarbaldehyde, tert-butyl n-4-formylcyclohexyl carbamate, trans-4-formyl-cyclohexyl-carbamic acid tert-butyl ester, tert-butyl trans-4-formylcyclohexyl carbamate, tert-butyl 4-formylcyclohexyl carbamate, trans-tert-butyl 4-formylcyclohexyl carbamate, tert-butyl 1r,4r-4-formylcyclohexylcarbamate, cis-4-boc-amino cyclohexanecarboxaldehyde
CC(C)(C)OC(=O)NC1CCC(CC1)C=O
227.304
227.3
cis-4-(Boc-amino)cyclohexanecarboxaldehyde
181308-56-5
MFCD03844599
GPDBIGSFXXKWQR-UHFFFAOYSA-N
tert-butyl N-(4-formylcyclohexyl)carbamate
11020615
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning