missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

cis-4-(Boc-amino)cyclohexaneacetic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H62722MD

 View more versions of this product

Catalog No. AAH62722MD



Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
cis-4-(Boc-amino)cyclohexaneacetic acid is generally used as a intermediate in chemical research.

Solubility
Slightly Soluble (1.1 g/L) (25°C).

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Specifications

cis-4-(Boc-amino)cyclohexaneacetic acid
C13H23NO4
2-cis-4-tert-butoxycarbonyl amino cyclohexyl acetic acid, trans-n-boc-4-aminocyclohexyl acetic acid, 2-trans-4-tert-butoxycarbonyl amino cyclohexyl acetic acid, boc-1,4-trans-acha-oh, cis-4-boc-amino cyclohexyl acetic acid, trans-n-boc-4-aminocyclohexyl aceticacid, 4-tert-butoxycarbonylamino-cyclohexyl-acetic acid, trans-4-tert-butoxycarbonylamino cyclohexylacetic acid, 1r,4r-4-tert-butoxycarbonyl amino cyclohexyl acetic acid, 1s,4s-4-tert-butoxycarbonyl amino cyclohexyl acetic acid
IHXBNSUFUFFBRL-UHFFFAOYSA-N
2-[4-[(2-methylpropan-2-yl)oxycarbonylamino]cyclohexyl]acetic acid
11253897
97%
327156-95-6
MFCD01862296,MFCD03844588
Slightly Soluble (1.1g/L) (25°C).
CC(C)(C)OC(=O)NC1CCC(CC(O)=O)CC1
257.33
257.32
250 mg

Chemical Identifiers

327156-95-6
257.33
IHXBNSUFUFFBRL-UHFFFAOYSA-N
11253897
C13H23NO4
MFCD01862296,MFCD03844588
2-cis-4-tert-butoxycarbonyl amino cyclohexyl acetic acid, trans-n-boc-4-aminocyclohexyl acetic acid, 2-trans-4-tert-butoxycarbonyl amino cyclohexyl acetic acid, boc-1,4-trans-acha-oh, cis-4-boc-amino cyclohexyl acetic acid, trans-n-boc-4-aminocyclohexyl aceticacid, 4-tert-butoxycarbonylamino-cyclohexyl-acetic acid, trans-4-tert-butoxycarbonylamino cyclohexylacetic acid, 1r,4r-4-tert-butoxycarbonyl amino cyclohexyl acetic acid, 1s,4s-4-tert-butoxycarbonyl amino cyclohexyl acetic acid
CC(C)(C)OC(=O)NC1CCC(CC(O)=O)CC1
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions