missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ cis-4-(Benzyloxycarbonylamino)cyclohexanecarboxylic acid, 97%

$31.76 - $373.19

Chemical Identifiers

CAS 67299-52-9
Molecular Formula C15H19NO4
Molecular Weight (g/mol) 277.32
MDL Number MFCD01880738
InChI Key ZVMICQYOGWAOSU-UHFFFAOYSA-N
Synonym trans-4-carbobenzoxyamino cyclohexanecarboxylic acid, trans-4-cbz-amino cyclohexanecarboxylic acid, 4-benzyloxycarbonylamino-cyclohexanecarboxylic acid, trans-4-benzyloxycarbonylaminocyclohexanecarboxylic acid, cis-4-benzyloxycarbonylaminocyclohexanecarboxylic acid, cis-4-benzyloxycarbonylaminocyclohexane-carboxylic acid, 4-benzyloxy carbonyl amino cyclohexanecarboxylic acid, 4-phenylmethoxycarbonylamino cyclohexane-1-carboxylic acid, trans-4-benzyloxy carbonyl amino cyclohexanecarboxylic acid, 4-benzyloxy carbonyl amino cyclohexanecarboxylic acid
PubChem CID 285247
IUPAC Name 4-(phenylmethoxycarbonylamino)cyclohexane-1-carboxylic acid
SMILES C1CC(CCC1C(=O)O)NC(=O)OCC2=CC=CC=C2
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62109MD View Documents Alfa Aesar™
H62109MD
250mg Each for $31.76
 
AAH6210903 View Documents Alfa Aesar™
H6210903
1g Each for $93.61
 
AAH6210906 View Documents Alfa Aesar™
H6210906
5g Each for $373.19
 
Description

Description

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

67299-52-9
277.32
ZVMICQYOGWAOSU-UHFFFAOYSA-N
285247
C1CC(CCC1C(=O)O)NC(=O)OCC2=CC=CC=C2
C15H19NO4
MFCD01880738
trans-4-carbobenzoxyamino cyclohexanecarboxylic acid, trans-4-cbz-amino cyclohexanecarboxylic acid, 4-benzyloxycarbonylamino-cyclohexanecarboxylic acid, trans-4-benzyloxycarbonylaminocyclohexanecarboxylic acid, cis-4-benzyloxycarbonylaminocyclohexanecarboxylic acid, cis-4-benzyloxycarbonylaminocyclohexane-carboxylic acid, 4-benzyloxy carbonyl amino cyclohexanecarboxylic acid, 4-phenylmethoxycarbonylamino cyclohexane-1-carboxylic acid, trans-4-benzyloxy carbonyl amino cyclohexanecarboxylic acid, 4-benzyloxy carbonyl amino cyclohexanecarboxylic acid
4-(phenylmethoxycarbonylamino)cyclohexane-1-carboxylic acid

Specifications

250mg
C15H19NO4
trans-4-carbobenzoxyamino cyclohexanecarboxylic acid, trans-4-cbz-amino cyclohexanecarboxylic acid, 4-benzyloxycarbonylamino-cyclohexanecarboxylic acid, trans-4-benzyloxycarbonylaminocyclohexanecarboxylic acid, cis-4-benzyloxycarbonylaminocyclohexanecarboxylic acid, cis-4-benzyloxycarbonylaminocyclohexane-carboxylic acid, 4-benzyloxy carbonyl amino cyclohexanecarboxylic acid, 4-phenylmethoxycarbonylamino cyclohexane-1-carboxylic acid, trans-4-benzyloxy carbonyl amino cyclohexanecarboxylic acid, 4-benzyloxy carbonyl amino cyclohexanecarboxylic acid
C1CC(CCC1C(=O)O)NC(=O)OCC2=CC=CC=C2
277.32
277.31
cis-4-(Benzyloxycarbonylamino)cyclohexanecarboxylic acid
67299-52-9
MFCD01880738
ZVMICQYOGWAOSU-UHFFFAOYSA-N
4-(phenylmethoxycarbonylamino)cyclohexane-1-carboxylic acid
285247
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning