missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
cis-4-(Benzyloxycarbonylamino)cyclohexanecarboxylic acid, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H6210906
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
SolubilitySoluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
cis-4-(Benzyloxycarbonylamino)cyclohexanecarboxylic acid | |
C15H19NO4 | |
trans-4-carbobenzoxyamino cyclohexanecarboxylic acid, trans-4-cbz-amino cyclohexanecarboxylic acid, 4-benzyloxycarbonylamino-cyclohexanecarboxylic acid, trans-4-benzyloxycarbonylaminocyclohexanecarboxylic acid, cis-4-benzyloxycarbonylaminocyclohexanecarboxylic acid, cis-4-benzyloxycarbonylaminocyclohexane-carboxylic acid, 4-benzyloxy carbonyl amino cyclohexanecarboxylic acid, 4-phenylmethoxycarbonylamino cyclohexane-1-carboxylic acid, trans-4-benzyloxy carbonyl amino cyclohexanecarboxylic acid | |
ZVMICQYOGWAOSU-UHFFFAOYSA-N | |
4-(phenylmethoxycarbonylamino)cyclohexane-1-carboxylic acid | |
285247 | |
97% |
67299-52-9 | |
MFCD01550737,MFCD01880738 | |
Soluble in water. | |
OC(=O)C1CCC(CC1)NC(=O)OCC1=CC=CC=C1 | |
277.32 | |
277.31 | |
5 g |
Chemical Identifiers
67299-52-9 | |
277.32 | |
ZVMICQYOGWAOSU-UHFFFAOYSA-N | |
285247 |
C15H19NO4 | |
MFCD01550737,MFCD01880738 | |
trans-4-carbobenzoxyamino cyclohexanecarboxylic acid, trans-4-cbz-amino cyclohexanecarboxylic acid, 4-benzyloxycarbonylamino-cyclohexanecarboxylic acid, trans-4-benzyloxycarbonylaminocyclohexanecarboxylic acid, cis-4-benzyloxycarbonylaminocyclohexanecarboxylic acid, cis-4-benzyloxycarbonylaminocyclohexane-carboxylic acid, 4-benzyloxy carbonyl amino cyclohexanecarboxylic acid, 4-phenylmethoxycarbonylamino cyclohexane-1-carboxylic acid, trans-4-benzyloxy carbonyl amino cyclohexanecarboxylic acid | |
OC(=O)C1CCC(CC1)NC(=O)OCC1=CC=CC=C1 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures