missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ cis-4-(Benzyloxycarbonylamino)cyclohexanecarboxaldehyde, 97%

$71.08 - $928.35

Chemical Identifiers

CAS 917022-26-5
Molecular Formula C15H19NO3
Molecular Weight (g/mol) 261.321
MDL Number MFCD03844593
InChI Key SOIRQISQWQDGSN-UHFFFAOYSA-N
Synonym benzyl 4-formylcyclohexylcarbamate, benzyl cis-4-formylcyclohexyl carbamate, benzyl cis-4-formylcyclohexylcarbamate, benzyl 4-formylcyclohexyl carbamate, trans-4-benzyloxycarbonylamino-cyclohexanecarbaldehyde, benzyl 1r,4r-4-formylcyclohexylcarbamate, trans-4-benzyloxycarbonylamino cyclohexanecarbaldehyde, benzyl n-4-formylcyclohexyl carbamate, benzyl4-formylcyclohexylcarbamate, soirqisqwqdgsn-mqmhxkeqsa-n
PubChem CID 21882542
IUPAC Name benzyl N-(4-formylcyclohexyl)carbamate
SMILES C1CC(CCC1C=O)NC(=O)OCC2=CC=CC=C2
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62546MD View Documents Alfa Aesar™
H62546MD
250mg Each for $71.08
 
AAH6254603 View Documents Alfa Aesar™
H6254603
1g Each for $211.27
 
AAH6254606 View Documents Alfa Aesar™
H6254606
5g Each for $928.35
 
Specifications

Chemical Identifiers

917022-26-5
261.321
SOIRQISQWQDGSN-UHFFFAOYSA-N
21882542
C1CC(CCC1C=O)NC(=O)OCC2=CC=CC=C2
C15H19NO3
MFCD03844593
benzyl 4-formylcyclohexylcarbamate, benzyl cis-4-formylcyclohexyl carbamate, benzyl cis-4-formylcyclohexylcarbamate, benzyl 4-formylcyclohexyl carbamate, trans-4-benzyloxycarbonylamino-cyclohexanecarbaldehyde, benzyl 1r,4r-4-formylcyclohexylcarbamate, trans-4-benzyloxycarbonylamino cyclohexanecarbaldehyde, benzyl n-4-formylcyclohexyl carbamate, benzyl4-formylcyclohexylcarbamate, soirqisqwqdgsn-mqmhxkeqsa-n
benzyl N-(4-formylcyclohexyl)carbamate

Specifications

250mg
C15H19NO3
benzyl 4-formylcyclohexylcarbamate, benzyl cis-4-formylcyclohexyl carbamate, benzyl cis-4-formylcyclohexylcarbamate, benzyl 4-formylcyclohexyl carbamate, trans-4-benzyloxycarbonylamino-cyclohexanecarbaldehyde, benzyl 1r,4r-4-formylcyclohexylcarbamate, trans-4-benzyloxycarbonylamino cyclohexanecarbaldehyde, benzyl n-4-formylcyclohexyl carbamate, benzyl4-formylcyclohexylcarbamate, soirqisqwqdgsn-mqmhxkeqsa-n
C1CC(CCC1C=O)NC(=O)OCC2=CC=CC=C2
261.321
261.31
cis-4-(Benzyloxycarbonylamino)cyclohexanecarboxaldehyde
917022-26-5
MFCD03844593
SOIRQISQWQDGSN-UHFFFAOYSA-N
benzyl N-(4-formylcyclohexyl)carbamate
21882542
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning