missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

cis-4-Amino-1-cyclohexanecarboxylic acid, 97%, ACROS Organics™

Chemical Identifiers

CAS 3685-23-2
Molecular Formula C7H13NO2
Molecular Weight (g/mol) 143.19
MDL Number MFCD00191730
InChI Key DRNGLYHKYPNTEA-UHFFFAOYSA-N
Synonym 1s,4s-4-aminocyclohexanecarboxylic acid, 4-aminocyclohexanecarboxylic acid, cis-4-amino-1-cyclohexanecarboxylic acid, cis-4-aminocyclohexane carboxylic acid, cis-4-aminocyclohexanecarboxylic acid, cis-4-aminocyclohexanecarboxylicacid, cyclohexanecarboxylic acid, 4-amino-, trans, trans-4-amino-cyclohexanecarboxylic acid, trans-4-aminocyclohexane carboxylic acid, trans-4-aminocyclohexanecarboxylic acid
PubChem CID 171560
IUPAC Name 4-aminocyclohexane-1-carboxylic acid
SMILES C1CC(CCC1C(=O)O)N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC368330010
SDS View Documents
Thermo Fisher Scientific
AC368330010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

3685-23-2
143.19
DRNGLYHKYPNTEA-UHFFFAOYSA-N
171560
C1CC(CCC1C(=O)O)N
C7H13NO2
MFCD00191730
1s,4s-4-aminocyclohexanecarboxylic acid, 4-aminocyclohexanecarboxylic acid, cis-4-amino-1-cyclohexanecarboxylic acid, cis-4-aminocyclohexane carboxylic acid, cis-4-aminocyclohexanecarboxylic acid, cis-4-aminocyclohexanecarboxylicacid, cyclohexanecarboxylic acid, 4-amino-, trans, trans-4-amino-cyclohexanecarboxylic acid, trans-4-aminocyclohexane carboxylic acid, trans-4-aminocyclohexanecarboxylic acid
4-aminocyclohexane-1-carboxylic acid

Specifications

>300°C
Glass bottle
278°C to 283°C
3685-23-2
C7H13NO2
14, IV, 981
Solubility in water: soluble
C1CC(CCC1C(=O)O)N
143.19
143.19
97%
Authentic
White
1g
97%
MFCD00191730
1s,4s-4-aminocyclohexanecarboxylic acid, 4-aminocyclohexanecarboxylic acid, cis-4-amino-1-cyclohexanecarboxylic acid, cis-4-aminocyclohexane carboxylic acid, cis-4-aminocyclohexanecarboxylic acid, cis-4-aminocyclohexanecarboxylicacid, cyclohexanecarboxylic acid, 4-amino-, trans, trans-4-amino-cyclohexanecarboxylic acid, trans-4-aminocyclohexane carboxylic acid, trans-4-aminocyclohexanecarboxylic acid
DRNGLYHKYPNTEA-UHFFFAOYSA-N
4-aminocyclohexane-1-carboxylic acid
171560
Powder
cis-4-Amino-1-cyclohexanecarboxylic acid, 97%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS