Please login to your online account to display your discounted pricing

Alfa Aesar™ cis-3-Nonen-1-ol, 97%

$44.59 - $44.59

Chemical Identifiers

Chemical Name or Material cis-3-Nonen-1-ol
CAS 10340-23-5
Molecular Formula C9H18O
Formula Weight 142.24
MDL Number MFCD00010314
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1784606

  SDS
alfa aesar™
A1784606
5g Each for $44.59

AAA1784614

  SDS
alfa aesar™
A1784614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material cis-3-Nonen-1-ol
Flash Point 95°C (203°F)
Boiling Point 107° to 109°C (20mmHg)
Quantity 5g
CAS 10340-23-5
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
TSCA TSCA
Assay 97%
Molecular Formula C9H18O
Formula Weight 142.24
MDL Number MFCD00010314

Applications
Used as flavoring agent for fresh, tropical, melon notes.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Store in a tightly closed container in a cool, dry, well-ventilated area. Store away from strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.

2° to 8°C