Please login to your online account to display your discounted pricing

Cis-3-hexene, 97%, ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  AC379630050

 View more versions of this product

Catalog No. AC379630050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Purity Grade Extra Pure
Physical Form Liquid
Formula Weight 84.16
Quantity 5g
Color Clear colorless to light yellow
Density 0.677g/mL
CAS 2097470
Chemical Name or Material cis-3-Hexene
Infrared Spectrum Authentic
Molecular Formula C6H12
MDL Number MFCD00085300
Assay Percent Range 97%
Refractive Index 1.3935 to 1.3955 (20°C, 589nm)
Linear Formula CH3CH2CH=CHCH2CH3

Specifications

Chemical Name or Material cis-3-Hexene
CAS 2097470
Purity Grade Extra Pure
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H12
Linear Formula CH3CH2CH=CHCH2CH3
Formula Weight 84.16
MDL Number MFCD00085300