missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cis-3-hexene, 97%, ACROS Organics™

Chemical Identifiers

CAS 2097470
Assay Percent Range 97%
Molecular Formula C6H12
Linear Formula CH3CH2CH=CHCH2CH3
Formula Weight 84.16
MDL Number MFCD00085300
Chemical Name or Material cis-3-Hexene
Assay ≥96.0%
Grade Extra Pure
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC379630050 SDS Thermo Fisher Scientific
AC379630050
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

2097470
C6H12
84.16
cis-3-Hexene
Extra Pure
97%
CH3CH2CH=CHCH2CH3
MFCD00085300
≥96.0%

Specifications

Clear colorless to light yellow
2097470
Authentic
97%
1.3935 to 1.3955 (20°C, 589nm)
84.16
cis-3-Hexene
Extra Pure
5g
0.677g/mL
Liquid
C6H12
CH3CH2CH=CHCH2CH3
MFCD00085300
≥96.0%