Learn More
cis-3-Hexen-1-ol, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1031318
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
cis-3-Hexen-1-ol | |
0.848 | |
44°C (111°F) | |
C6H12O | |
MFCD00063217 | |
UN1987 | |
14,4700 | |
UFLHIIWVXFIJGU-ARJAWSKDSA-N | |
(Z)-hex-3-en-1-ol | |
5281167 | |
100.16 |
928-96-1 | |
156°C to 157°C | |
Pleasant | |
1.44 | |
50 g | |
1719712 | |
z-hex-3-en-1-ol, cis-3-hexen-1-ol, cis-3-hexenol, leaf alcohol, blatteralkohol, z-3-hexenol, z-3-hexen-1-ol, 3-hexenol, 3z-hex-3-en-1-ol, 3-hexen-1-ol, z | |
CC\C=C/CCO | |
100.16 | |
CHEBI:28857 | |
98% |
Chemical Identifiers
928-96-1 | |
100.16 | |
UFLHIIWVXFIJGU-ARJAWSKDSA-N | |
5281167 | |
CC\C=C/CCO |
C6H12O | |
MFCD00063217 | |
z-hex-3-en-1-ol, cis-3-hexen-1-ol, cis-3-hexenol, leaf alcohol, blatteralkohol, z-3-hexenol, z-3-hexen-1-ol, 3-hexenol, 3z-hex-3-en-1-ol, 3-hexen-1-ol, z | |
CHEBI:28857 |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P370+P378q-P501c
H226-H319
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber : 213-192-8
RTECSNumber : MP8400000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only