Please login to your online account to display your discounted pricing

Alfa Aesar™ cis-3-Hepten-1-ol, 97%

$82.97 - $289.38

Chemical Identifiers

Chemical Name or Material cis-3-Hepten-1-ol
CAS 1708-81-2
Molecular Formula C7H14O
Formula Weight 114.18
MDL Number MFCD00274760
Beilstein 1719803
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1917206

 
alfa aesar™
A1917206
5g Each for $82.97

AAA1917214

 
alfa aesar™
A1917214
25g Each for $289.38
Description & Specifications

Specifications

Formula Weight 114.18
Chemical Name or Material cis-3-Hepten-1-ol
TSCA TSCA
Assay 97%
Molecular Formula C7H14O
MDL Number MFCD00274760
CAS 1708-81-2
Beilstein 1719803
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Boiling Point 92°C (60mmHg)
Quantity 5g
Flash Point 68°C (154°F)
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
cis-3-Hepten-1-ol is used in the analysis of hop-derived terpenoids in beer. Imparts a fresh flower garden odor so used in flavoring agent.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.