missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ cis-3-Chloroacrylic acid, 98+%

Manufacturer:  Alfa Aesar™ A1479806

 View more versions of this product

Catalog No. AAA1479806



Specifications

Specifications

cis-3-Chloroacrylic acid
5g
C3H3ClO2
UN3261
cis-3-chloroacrylic acid, z-3-chloroacrylic acid, cis-beta-chloroacrylic acid, 2-propenoic acid, 3-chloro-, z, cis-3-chloropropenoic acid, 2z-3-chloroacrylic acid, 2-propenoic acid, 3-chloro-, 2z, cis-.beta.-chloroacrylic acid, z-3-chloroprop-2-enoic acid, acrylic acid, 3-chloro-, z-8ci
C(=CCl)C(=O)O
106.505
CHEBI:27397
≥98%
60°C to 62°C
1609-93-4
MFCD00004368
1720278
MHMUCYJKZUZMNJ-UPHRSURJSA-N
(Z)-3-chloroprop-2-enoic acid
643794
106.51

Chemical Identifiers

1609-93-4
106.505
MHMUCYJKZUZMNJ-UPHRSURJSA-N
643794
(Z)-3-chloroprop-2-enoic acid
C3H3ClO2
MFCD00004368
cis-3-chloroacrylic acid, z-3-chloroacrylic acid, cis-beta-chloroacrylic acid, 2-propenoic acid, 3-chloro-, z, cis-3-chloropropenoic acid, 2z-3-chloroacrylic acid, 2-propenoic acid, 3-chloro-, 2z, cis-.beta.-chloroacrylic acid, z-3-chloroprop-2-enoic acid, acrylic acid, 3-chloro-, z-8ci
CHEBI:27397
C(=CCl)C(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

Recommended Storage : 2° to 8°C

Documents