missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
cis-3-(Boc-amino)cyclobutanemethanol, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H6268603
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
cis-3-(Boc-amino)cyclobutanemethanol | |
C10H19NO3 | |
tert-butyl n-3-hydroxymethyl cyclobutyl carbamate, tert-butyl cis-3-hydroxymethyl cyclobutyl carbamate, 3-boc-amino-cyclobutanemethanol, tert-butyl cis-3-hydroxymethylcyclobutylcarbamate, tert-butyl trans-3-hydroxymethylcyclobutylcarbamate, tert-butyl 3-hydroxymethyl cyclobutyl carbamate, cis-1-boc-amino-3-hydroxymethyl cyclobutane, trans-1-boc-amino-3-hydroxymethyl cyclobutane, tert-butyl trans-3-hydroxymethyl cyclobutyl carbamate, cis-3-boc-amino-cyclobutanemethanol | |
CC(C)(C)OC(=O)NC1CC(C1)CO | |
201.266 | |
201.26 | |
1 g |
142733-64-0 | |
MFCD08436074 | |
PCPNTJQMXAHNOA-UHFFFAOYSA-N | |
tert-butyl N-[3-(hydroxymethyl)cyclobutyl]carbamate | |
22408683 | |
97% |
Chemical Identifiers
142733-64-0 | |
201.266 | |
PCPNTJQMXAHNOA-UHFFFAOYSA-N | |
22408683 | |
CC(C)(C)OC(=O)NC1CC(C1)CO |
C10H19NO3 | |
MFCD08436074 | |
tert-butyl n-3-hydroxymethyl cyclobutyl carbamate, tert-butyl cis-3-hydroxymethyl cyclobutyl carbamate, 3-boc-amino-cyclobutanemethanol, tert-butyl cis-3-hydroxymethylcyclobutylcarbamate, tert-butyl trans-3-hydroxymethylcyclobutylcarbamate, tert-butyl 3-hydroxymethyl cyclobutyl carbamate, cis-1-boc-amino-3-hydroxymethyl cyclobutane, trans-1-boc-amino-3-hydroxymethyl cyclobutane, tert-butyl trans-3-hydroxymethyl cyclobutyl carbamate, cis-3-boc-amino-cyclobutanemethanol | |
tert-butyl N-[3-(hydroxymethyl)cyclobutyl]carbamate |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures