missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ cis-3-(Benzyloxycarbonylamino)cyclohexanemethanol, 97%

Chemical Identifiers

CAS 1931962-56-9
Molecular Formula C15H21NO3
MDL Number MFCD09787485
Synonym Benzyl cis-3-hydroxymethylcyclohexylcarbamate; cis-3-(Benzyloxycarbonylamino)-1-hydroxymethylcyclohexane
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62094MD View Documents Alfa Aesar™
H62094MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6209403 View Documents Alfa Aesar™
H6209403
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

1931962-56-9
C15H21NO3
MFCD09787485
Benzyl cis-3-hydroxymethylcyclohexylcarbamate; cis-3-(Benzyloxycarbonylamino)-1-hydroxymethylcyclohexane

Specifications

250mg
C15H21NO3
Benzyl cis-3-hydroxymethylcyclohexylcarbamate; cis-3-(Benzyloxycarbonylamino)-1-hydroxymethylcyclohexane
97%
1931962-56-9
MFCD09787485
263.33
cis-3-(Benzyloxycarbonylamino)cyclohexanemethanol