Please login to your online account to display your discounted pricing

Alfa Aesar™ cis-3-Acetyl-2,2-dimethylcyclobutaneacetic acid, 97%

Chemical Identifiers

Chemical Name or Material cis-3-Acetyl-2,2-dimethylcyclobutaneacetic acid
CAS 64396-97-0
Formula Weight 184.23
MDL Number MFCD09864988
Synonym 2-[(1S,3S)-3-Acetyl-2,2-dimethylcyclobutyl]acetic acid; cis-Pinonic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6247503

 
alfa aesar™
H6247503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62475MD

 
alfa aesar™
H62475MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 104° to 107°C
Formula Weight 184.23
Quantity 1g
Physical Form Solid
Assay 97%
CAS 64396-97-0
Chemical Name or Material cis-3-Acetyl-2,2-dimethylcyclobutaneacetic acid
Synonym 2-[(1S,3S)-3-Acetyl-2,2-dimethylcyclobutyl]acetic acid; cis-Pinonic acid
MDL Number MFCD09864988