Please login to your online account to display your discounted pricing

cis-2-(tert-Butoxycarbonylamino)-1-cyclopentanecarboxylic acid, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC299580010

 View more versions of this product

Catalog No. AC299580010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material cis-2-(tert-Butoxycarbonylamino)-1-cyclopentanecarboxylic acid
CAS 136315-70-3
Infrared Spectrum Authentic
Physical Form Fine Crystalline Powder
Assay ≥97.5%
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C11H19NO4
Formula Weight 229.28
Color White to almost white
Quantity 1g

Specifications

Chemical Name or Material cis-2-(tert-Butoxycarbonylamino)-1-cyclopentanecarboxylic acid
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C11H19NO4
Formula Weight 229.28