missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

cis-2-Methoxycinnamic Acid, TCI America™

$145.14 - $412.52

Chemical Identifiers

CAS 14737-91-8
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.187
MDL Number MFCD00004375
InChI Key FEGVSPGUHMGGBO-SREVYHEPSA-N
Synonym cis-O-Methyl-o-coumaric Acid, cis-O-Methyl-o-cumaric Acid
PubChem CID 1622530
IUPAC Name (Z)-3-(2-methoxyphenyl)prop-2-enoic acid
SMILES COC1=CC=CC=C1C=CC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M07615G View Documents TCI America
M07615G
5g Each for $145.14
 
M076125G View Documents TCI America
M076125G
25g Each for $412.52
 
Specifications

Chemical Identifiers

14737-91-8
178.187
FEGVSPGUHMGGBO-SREVYHEPSA-N
1622530
COC1=CC=CC=C1C=CC(=O)O
C10H10O3
MFCD00004375
cis-O-Methyl-o-coumaric Acid, cis-O-Methyl-o-cumaric Acid
(Z)-3-(2-methoxyphenyl)prop-2-enoic acid

Specifications

White-Yellow
5g
C10H10O3
cis-O-Methyl-o-coumaric Acid, cis-O-Methyl-o-cumaric Acid
COC1=CC=CC=C1C=CC(=O)O
178.187
178.19
≥98.0% (T)
94°C
14737-91-8
MFCD00004375
FEGVSPGUHMGGBO-SREVYHEPSA-N
(Z)-3-(2-methoxyphenyl)prop-2-enoic acid
1622530
Crystal-Powder at 20°C
cis-2-Methoxycinnamic Acid
Safety and Handling

Safety and Handling

RTECSNumber : GD9720000

TSCA : No