missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
cis-2-Methoxycinnamic Acid 98.0+%, TCI America™
Supplier: TCI America M076125G
Specifications
cis-2-Methoxycinnamic Acid | |
94°C | |
C10H10O3 | |
25 g | |
FEGVSPGUHMGGBO-SREVYHEPSA-N | |
(Z)-3-(2-methoxyphenyl)prop-2-enoic acid | |
1622530 | |
≥98.0% (T) |
14737-91-8 | |
White-Yellow | |
MFCD00004375 | |
cis-O-Methyl-o-coumaric Acid, cis-O-Methyl-o-cumaric Acid | |
COC1=CC=CC=C1C=CC(=O)O | |
178.187 | |
178.19 | |
Crystalline Powder |
Chemical Identifiers
14737-91-8 | |
178.187 | |
FEGVSPGUHMGGBO-SREVYHEPSA-N | |
1622530 | |
COC1=CC=CC=C1C=CC(=O)O |
C10H10O3 | |
MFCD00004375 | |
cis-O-Methyl-o-coumaric Acid, cis-O-Methyl-o-cumaric Acid | |
(Z)-3-(2-methoxyphenyl)prop-2-enoic acid |
Safety and Handling
RTECSNumber : GD9720000
TSCA : No