Please login to your online account to display your discounted pricing

cis-2-Hydroxymethyl-1-cyclohexylamine hydrochloride, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material cis-2-Hydroxymethyl-1-cyclohexylamine hydrochloride
Name Note 99%
CAS 5691-37-2
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H15NO·HCl
Formula Weight 165.66
MDL Number MFCD00143988
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC265860010

  SDS
fisher scientific acros
265860010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Formula Weight 165.66
Physical Form Crystalline Powder
Quantity 1g
Color White to light beige
Melting Point 131° to 135°C
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material cis-2-Hydroxymethyl-1-cyclohexylamine hydrochloride
Name Note 99%
MDL Number MFCD00143988
Molecular Formula C7H15NO·HCl
CAS 5691-37-2
Assay Percent Range 99%