missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
cis-2-Hydroxymethyl-1-cyclohexylamine hydrochloride, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 265860010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
cis-2-Hydroxymethyl-1-cyclohexylamine hydrochloride | |
5691-37-2 | |
98.5% min. (Argentometry) | |
C7H15NO·HCl | |
1g | |
Solubility in water: soluble. Other solubilities: soluble in methanol | |
C1CCC(C(C1)CO)N.Cl | |
165.66 | |
165.66 |
99% | |
Authentic | |
Glass bottle | |
MFCD00143988 | |
cis-2-hydroxymethyl-1-cyclohexylamine hydrochloride, cis-2-aminocyclohexyl methanol hydrochloride, 1s,2r-2-aminocyclohexyl methanol hydrochloride, +/-cis-2-hydroxymethyl-1-cyclohexylamine hydrochloride, racemic-cis-2-hydroxymethyl-1-cyclohexylamine hydrochloride | |
ORPKUPNVXFLMRG-ZJLYAJKPSA-N | |
[(1S,2R)-2-aminocyclohexyl]methanol;hydrochloride | |
23545866 | |
99% |
RUO â Research Use Only