Please login to your online account to display your discounted pricing

cis-2-Hydroxy-1-cyclohexanecarboxamide, 98+%, ACROS Organics™


Quantity 500mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  277715000

 View more versions of this product

Catalog No. AC277715000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.0%
Physical Form Crystalline Powder and Chunks
Formula Weight 143.19
Quantity 500mg
Color White
Melting Point 115° to 119°C
Packaging Glass bottle
CAS 73045-98-4
Synonym cis-Hexahydrosalicylamide
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Chemical Name or Material cis-2-Hydroxy-1-cyclohexanecarboxamide
Name Note 98+%
Assay Percent Range 98+%
Molecular Formula C7H13NO2
MDL Number MFCD00143985

Specifications

Chemical Name or Material cis-2-Hydroxy-1-cyclohexanecarboxamide
Name Note 98+%
CAS 73045-98-4
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C7H13NO2
Formula Weight 143.19
MDL Number MFCD00143985
Synonym cis-Hexahydrosalicylamide
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents