missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

cis-2-Hydroxy-1-cyclohexanecarboxamide, 98+%, ACROS Organics™

$209.99 - $209.99

Chemical Identifiers

CAS 73045-98-4
Assay Percent Range 98+%
Molecular Formula C7H13NO2
Formula Weight 143.19
MDL Number MFCD00143985
Synonym cis-Hexahydrosalicylamide
Chemical Name or Material cis-2-Hydroxy-1-cyclohexanecarboxamide, 98+%
Assay ≥98.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC277715000 SDS Thermo Fisher Scientific
277715000
Glass bottle 500mg Each for $209.99
Specifications Description & Specifications

Chemical Identifiers

73045-98-4
C7H13NO2
MFCD00143985
cis-2-Hydroxy-1-cyclohexanecarboxamide, 98+%
98+%
143.19
cis-Hexahydrosalicylamide
≥98.0%

Specifications

White
500mg
Authentic
Crystalline Powder and Chunks
C7H13NO2
MFCD00143985
cis-2-Hydroxy-1-cyclohexanecarboxamide, 98+%
115° to 119°C
73045-98-4
Glass bottle
98+%
143.19
cis-Hexahydrosalicylamide
≥98.0%

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning