Please login to your online account to display your discounted pricing

Alfa Aesar™ cis-2-Hexene, 96%

$55.89 - $55.89

Chemical Identifiers

Chemical Name or Material cis-2-Hexene
CAS 7688-21-3
Molecular Formula C6H12
Formula Weight 84.16
MDL Number MFCD00066520
UN Number UN3295
Beilstein 1719019
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5350606

  SDS
alfa aesar™
H5350606
5g Each for $55.89
Description & Specifications

Specifications

Formula Weight 84.16
Chemical Name or Material cis-2-Hexene
Assay 96%
Molecular Formula C6H12
MDL Number MFCD00066520
CAS 7688-21-3
UN Number UN3295
Beilstein 1719019
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Boiling Point 68° to 70°C
Quantity 5g
Flash Point −25°C (−13°F)
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.

Applications
cis-2-Hexene is used as an organic chemical synthesis intermediate.

Solubility
Immiscible in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.