missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ cis-2-Hexene, 96%

Manufacturer:  Alfa Aesar™ H5350614

 View more versions of this product

Catalog No. AAH5350614



Description

Description

Applications
cis-2-Hexene is used as an organic chemical synthesis intermediate.

Solubility
Immiscible in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
Specifications

Specifications

cis-2-Hexene
68°C to 70°C
7688-21-3
MFCD00066520
1719019
RYPKRALMXUUNKS-HYXAFXHYSA-N
(Z)-hex-2-ene
643835
96%
−25°C (−13°F)
25g
C6H12
UN3295
2-hexene z
CCCC=CC
84.162
84.16

Chemical Identifiers

7688-21-3
84.162
RYPKRALMXUUNKS-HYXAFXHYSA-N
643835
CCCC=CC
C6H12
MFCD00066520
2-hexene z
(Z)-hex-2-ene
Safety and Handling

Safety and Handling

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.

Documents