Please login to your online account to display your discounted pricing

cis-2-Hexene 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material cis-2-Hexene
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C6H12
Linear Formula CH3CH=CH(CH2)2CH3
Formula Weight 84.16
MDL Number MFCD00066520
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC389940010

  SDS
thermo fisher scientific
AC389940010
Glass bottle 1mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC389940050

  SDS
thermo fisher scientific
AC389940050
Glass bottle 5mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Formula Weight 84.16
Physical Form Liquid
Quantity 1mL
Color Clear colorless
Density 0.687g/mL
CAS 7688-21-3
Linear Formula CH3CH=CH(CH2)2CH3
Molecular Formula C6H12
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Highly flammable liquid and vapor.
MDL Number MFCD00066520
Infrared Spectrum Authentic
Refractive Index 1.3960 to 1.3980 (20°C, 589nm)
Assay Percent Range 95%
Packaging Glass bottle
Chemical Name or Material cis-2-Hexene
Health Hazard 1 GHS Signal Word: Danger
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Highly flammable liquid and vapor.