Please login to your online account to display your discounted pricing

Alfa Aesar™ cis-2-Hexen-1-ol, 94%, remainder mainly trans isomer

Chemical Identifiers

Chemical Name or Material cis-2-Hexen-1-ol
CAS 928-94-9
Molecular Formula C6H12O
Formula Weight 100.16
MDL Number MFCD00063209
UN Number UN1987
Beilstein 1719708
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1373406

  SDS
alfa aesar™
A1373406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1373414

  SDS
alfa aesar™
A1373414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1373422

  SDS
alfa aesar™
A1373422
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 100.16
Chemical Name or Material cis-2-Hexen-1-ol
TSCA TSCA
Assay 94%
Assay Percent Notes remainder mainly trans isomer
Molecular Formula C6H12O
MDL Number MFCD00063209
CAS 928-94-9
UN Number UN1987
Beilstein 1719708
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H319
Flammable liquid and vapour.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
RTECS Number MP8395000
Flash Point 54°C (129°F)
Boiling Point 158°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H319
Flammable liquid and vapour.
Causes serious eye irritation.