Please login to your online account to display your discounted pricing

Alfa Aesar™ cis-2-(Boc-amino)cyclohexanecarboxylic acid, 97%

Chemical Identifiers

Chemical Name or Material cis-2-(Boc-amino)cyclohexanecarboxylic acid
CAS 63216-49-9
Formula Weight 243.29
MDL Number MFCD01863244
Synonym cis-2-tert-Butoxycarbonylaminocyclohexanecarboxylic acid; Boc-1,2-cis-ACHC-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6260003

 
alfa aesar™
H6260003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6260006

 
alfa aesar™
H6260006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62600MD

 
alfa aesar™
H62600MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 243.29
Melting Point 127° to 131°C
Quantity 1g
Chemical Name or Material cis-2-(Boc-amino)cyclohexanecarboxylic acid
Physical Form Solid
Assay 97%
MDL Number MFCD01863244
CAS 63216-49-9
Synonym cis-2-tert-Butoxycarbonylaminocyclohexanecarboxylic acid; Boc-1,2-cis-ACHC-OH