missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ cis-2-(Benzyloxycarbonylamino)cyclohexanecarboxylic acid, 97%

Manufacturer:  Alfa Aesar™ H6254803

 View more versions of this product

Catalog No. AAH6254803



Specifications

Specifications

cis-2-(Benzyloxycarbonylamino)cyclohexanecarboxylic acid
54867-08-2
MFCD01863245
RPJMLWMATNCSIS-QWHCGFSZSA-N
(1S,2R)-2-(phenylmethoxycarbonylamino)cyclohexane-1-carboxylic acid
18599068
Solid
1g
C15H19NO4
1r,2s-2-benzyloxy carbonyl amino cyclohexane-1-carboxylic acid
C1CCC(C(C1)C(=O)O)NC(=O)OCC2=CC=CC=C2
277.32
277.31
97%

Chemical Identifiers

54867-08-2
277.32
RPJMLWMATNCSIS-QWHCGFSZSA-N
18599068
C1CCC(C(C1)C(=O)O)NC(=O)OCC2=CC=CC=C2
C15H19NO4
MFCD01863245
1r,2s-2-benzyloxy carbonyl amino cyclohexane-1-carboxylic acid
(1S,2R)-2-(phenylmethoxycarbonylamino)cyclohexane-1-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents