missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ cis-2-(Benzyloxycarbonylamino)cyclobutanecarboxylic acid, 97%

$350.07 - $1,116.50

Chemical Identifiers

CAS 685508-28-5
Molecular Formula C13H15NO4
Molecular Weight (g/mol) 249.266
MDL Number MFCD03844615
InChI Key ZNRUEEQIUVKKBL-MNOVXSKESA-N
Synonym cis-2-benzyloxycarbonylaminocyclobutane-carboxylic acid, cis-2-bocamino cyclobutanecarboxylic acid, cis-2-cbz-amino cyclobutane-1-carboxylic acid, 2beta-benzyloxycarbonylamino cyclobutane-1beta-carboxylic acid, 1r,2s-2-benzyloxy carbonyl amino cyclobutane-1-carboxylic acid
PubChem CID 68498468
IUPAC Name (1R,2S)-2-(phenylmethoxycarbonylamino)cyclobutane-1-carboxylic acid
SMILES C1CC(C1C(=O)O)NC(=O)OCC2=CC=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62329MD View Documents Alfa Aesar™
H62329MD
250mg Each for $350.07
 
AAH6232903 View Documents Alfa Aesar™
H6232903
1g Each for $1,116.50
 
Description

Description

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
Specifications

Chemical Identifiers

685508-28-5
249.266
ZNRUEEQIUVKKBL-MNOVXSKESA-N
68498468
C1CC(C1C(=O)O)NC(=O)OCC2=CC=CC=C2
C13H15NO4
MFCD03844615
cis-2-benzyloxycarbonylaminocyclobutane-carboxylic acid, cis-2-bocamino cyclobutanecarboxylic acid, cis-2-cbz-amino cyclobutane-1-carboxylic acid, 2beta-benzyloxycarbonylamino cyclobutane-1beta-carboxylic acid, 1r,2s-2-benzyloxy carbonyl amino cyclobutane-1-carboxylic acid
(1R,2S)-2-(phenylmethoxycarbonylamino)cyclobutane-1-carboxylic acid

Specifications

250mg
C13H15NO4
cis-2-benzyloxycarbonylaminocyclobutane-carboxylic acid, cis-2-bocamino cyclobutanecarboxylic acid, cis-2-cbz-amino cyclobutane-1-carboxylic acid, 2beta-benzyloxycarbonylamino cyclobutane-1beta-carboxylic acid, 1r,2s-2-benzyloxy carbonyl amino cyclobutane-1-carboxylic acid
C1CC(C1C(=O)O)NC(=O)OCC2=CC=CC=C2
249.266
249.26
cis-2-(Benzyloxycarbonylamino)cyclobutanecarboxylic acid
685508-28-5
MFCD03844615
ZNRUEEQIUVKKBL-MNOVXSKESA-N
(1R,2S)-2-(phenylmethoxycarbonylamino)cyclobutane-1-carboxylic acid
68498468
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning