missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(+)-cis-2-Benzylaminocyclohexanemethanol 98.0+%, TCI America™
$220.65 - $680.19
Chemical Identifiers
CAS | 71581-92-5 |
---|---|
Molecular Formula | C14H21NO |
Molecular Weight (g/mol) | 219.328 |
MDL Number | MFCD00151395 |
InChI Key | BRQFIORUNWWNBM-KBPBESRZSA-N |
Synonym | 1r,2s-2-benzylamino cyclohexyl methanol, +-cis-2-benzylaminocyclohexanemethanol, cis-1r,2s-+-2-benzylamino cyclohexanemethanol, --cis-2-benzylaminocyclohexanemethanol, cis-1r,2s-+-2-benzylaminocyclohexanemethanol, pubchem7939, cis-2-benzylamino cyclohexylmethanol, 2-benzylamino cyclohexyl methanol #, cis-2-benzylamino cyclohexyl methanol |
PubChem CID | 853687 |
IUPAC Name | [(1R,2S)-2-(benzylamino)cyclohexyl]methanol |
SMILES | C1CCC(C(C1)CO)NCC2=CC=CC=C2 |
Chemical Identifiers
71581-92-5 | |
219.328 | |
BRQFIORUNWWNBM-KBPBESRZSA-N | |
853687 | |
C1CCC(C(C1)CO)NCC2=CC=CC=C2 |
C14H21NO | |
MFCD00151395 | |
1r,2s-2-benzylamino cyclohexyl methanol, +-cis-2-benzylaminocyclohexanemethanol, cis-1r,2s-+-2-benzylamino cyclohexanemethanol, --cis-2-benzylaminocyclohexanemethanol, cis-1r,2s-+-2-benzylaminocyclohexanemethanol, pubchem7939, cis-2-benzylamino cyclohexylmethanol, 2-benzylamino cyclohexyl methanol #, cis-2-benzylamino cyclohexyl methanol | |
[(1R,2S)-2-(benzylamino)cyclohexyl]methanol |
Specifications
71581-92-5 | |
C14H21NO | |
1 g | |
BRQFIORUNWWNBM-KBPBESRZSA-N | |
[(1R,2S)-2-(benzylamino)cyclohexyl]methanol | |
853687 | |
≥98.0% (T) | |
(+)-cis-2-Benzylaminocyclohexanemethanol |
68°C | |
MFCD00151395 | |
1r,2s-2-benzylamino cyclohexyl methanol, +-cis-2-benzylaminocyclohexanemethanol, cis-1r,2s-+-2-benzylamino cyclohexanemethanol, --cis-2-benzylaminocyclohexanemethanol, cis-1r,2s-+-2-benzylaminocyclohexanemethanol, pubchem7939, cis-2-benzylamino cyclohexylmethanol, 2-benzylamino cyclohexyl methanol #, cis-2-benzylamino cyclohexyl methanol | |
C1CCC(C(C1)CO)NCC2=CC=CC=C2 | |
219.328 | |
219.33 | |
Crystalline Powder |
Safety and Handling
TSCA : No