missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(-)-cis-2-Benzylaminocyclohexanemethanol, TCI America™

$192.50 - $575.98

Chemical Identifiers

CAS 71581-93-6
Molecular Formula C14H21NO
Molecular Weight (g/mol) 219.328
MDL Number MFCD00151396
InChI Key BRQFIORUNWWNBM-ZIAGYGMSSA-N
Synonym 1r,2s-n-benzyl-2-hydroxymethyl cyclohexan-1-aminium
PubChem CID 853690
IUPAC Name [(1S,2R)-2-(benzylamino)cyclohexyl]methanol
SMILES C1CCC(C(C1)CO)NCC2=CC=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B11181G View Documents TCI America
B11181G
1g Each for $192.50
 
B11185G View Documents TCI America
B11185G
5g Each for $575.98
 
Specifications

Chemical Identifiers

71581-93-6
219.328
BRQFIORUNWWNBM-ZIAGYGMSSA-N
853690
C1CCC(C(C1)CO)NCC2=CC=CC=C2
C14H21NO
MFCD00151396
1r,2s-n-benzyl-2-hydroxymethyl cyclohexan-1-aminium
[(1S,2R)-2-(benzylamino)cyclohexyl]methanol

Specifications

White-Yellow
1g
C14H21NO
1r,2s-n-benzyl-2-hydroxymethyl cyclohexan-1-aminium
C1CCC(C(C1)CO)NCC2=CC=CC=C2
219.328
219.33
≥98.0% (T)
68°C
71581-93-6
MFCD00151396
BRQFIORUNWWNBM-ZIAGYGMSSA-N
[(1S,2R)-2-(benzylamino)cyclohexyl]methanol
853690
Crystal-Powder at 20°C
(-)-cis-2-Benzylaminocyclohexanemethanol
Safety and Handling

Safety and Handling

TSCA : No