missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

cis-2-Aminomethyl-1-cyclohexanol hydrochloride, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 265882500

 View more versions of this product

Catalog No. AC265882500



Description

Description

Specifications

Specifications

cis-2-Aminomethyl-1-cyclohexanol hydrochloride
Glass bottle
182.0°C to 185.0°C
99%
98.5% min. (Argentometry)
MFCD00143984
Solubility in water: soluble. Other solubilities: soluble in methanol
C1CCC(C(C1)CN)O.Cl
165.66
165.66
99%
Authentic
White
250mg
24947-68-0
C7H15NO·HCl
1s,2s-2-hydroxycyclohexyl methanaminium
UVYXWKUCIQHOCP-LEUCUCNGSA-N
(1S,2S)-2-(aminomethyl)cyclohexan-1-ol;hydrochloride
2724656
Crystalline Powder

Chemical Identifiers

24947-68-0
165.66
UVYXWKUCIQHOCP-LEUCUCNGSA-N
2724656
C1CCC(C(C1)CN)O.Cl
C7H15NO·HCl
MFCD00143984
1s,2s-2-hydroxycyclohexyl methanaminium
(1S,2S)-2-(aminomethyl)cyclohexan-1-ol;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable solid.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents