missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
cis-2-Amino-1-cyclopentanecarboxylic acid, 98+%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 277745000
Specifications
cis-2-Amino-1-cyclopentanecarboxylic acid | |
218.0°C to 220.0°C | |
98+% | |
98+% | |
MFCD00143982 | |
JWYOAMOZLZXDER-UHNVWZDZSA-N | |
(1R,2S)-2-aminocyclopentane-1-carboxylic acid | |
73305 | |
98+% |
Glass bottle | |
500mg | |
37910-65-9 | |
C6H11NO2 | |
+/--cispentacin, --1r,2s-cispentacin, --cispentacin, 1r,2s-2-aminocyclopentane-1-carboxylic acid, 1r,2s-2-aminocyclopentanecarboxylic acid, antibiotic fr 109615, cis-2-amino-1-cyclopentanecarboxylic acid, cis-pentacin, cispentacin, cispentacin,- | |
C1CC(C(C1)N)C(=O)O | |
129.159 | |
129.16 |
Chemical Identifiers
37910-65-9 | |
129.159 | |
JWYOAMOZLZXDER-UHNVWZDZSA-N | |
73305 | |
C1CC(C(C1)N)C(=O)O |
C6H11NO2 | |
MFCD00143982 | |
+/--cispentacin, --1r,2s-cispentacin, --cispentacin, 1r,2s-2-aminocyclopentane-1-carboxylic acid, 1r,2s-2-aminocyclopentanecarboxylic acid, antibiotic fr 109615, cis-2-amino-1-cyclopentanecarboxylic acid, cis-pentacin, cispentacin, cispentacin,- | |
(1R,2S)-2-aminocyclopentane-1-carboxylic acid |