Please login to your online account to display your discounted pricing

cis-2-Amino-1-cyclohexanecarboxylic acid, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material cis-2-Amino-1-cyclohexanecarboxylic acid
Name Note 98%
CAS 5691-20-3
Assay ≥97.5% (on dry substance)
Assay Percent Range 98%
Molecular Formula C7H13NO2
Formula Weight 143.19
MDL Number MFCD00143980
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC265625000

  SDS
fisher scientific acros
AC265625000
Glass bottle 500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Assay ≥97.5% (on dry substance)
Formula Weight 143.19
Quantity 500mg
Color Almost white to light beige
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Name Note 98%
Chemical Name or Material cis-2-Amino-1-cyclohexanecarboxylic acid
CAS 5691-20-3
Assay Percent Range 98%
Molecular Formula C7H13NO2
MDL Number MFCD00143980
Water 6% max. (K.F.)
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.