missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

cis-2-Amino-1-cyclohexanecarboxamide, 98+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC277770010

 View more versions of this product

Catalog No. AC277770010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

cis-2-Amino-1-cyclohexanecarboxamide
Glass bottle
124°C to 127°C
98+%
98+%
MFCD00143979
Solubility in water: .. Other solubilities: soluble in most common organic solvents
C1CCC(C(C1)C(=O)N)N
142.2
142.2
98+%
Authentic
White
1g
115014-77-2
C7H14N2O
1s-2beta-aminocyclohexane-1alpha-carboxamide, cyclohexanecarboxamide,2-amino-, 1r,2s-rel
DXNKJRKPVQVDJW-WDSKDSINSA-N
(1S,2S)-2-aminocyclohexane-1-carboxamide
30069041
Crystalline Powder

Chemical Identifiers

115014-77-2
142.2
DXNKJRKPVQVDJW-WDSKDSINSA-N
30069041
C1CCC(C(C1)C(=O)N)N
C7H14N2O
MFCD00143979
1s-2beta-aminocyclohexane-1alpha-carboxamide, cyclohexanecarboxamide,2-amino-, 1r,2s-rel
(1S,2S)-2-aminocyclohexane-1-carboxamide
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents